Supelco® Drug and metabolite standards, naltrexone solution, (CRM)
Catalog # N-007-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:1,0 mg/ml in methanol
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:16590-41-3
- Description:Naltrexone solution
- Description:Naltrexone solution
- Pk Info:ampoule of 1 ml