Supelco® Drug and metabolite standards, mycophenolic acid-d3 solution, (CRM)
Catalog # M-137-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 µg/ml in acetonitrile
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:1185242-90-3
- Description:Mycophenolic acid-d3 solution
- Description:Mycophenolic acid-d3 solution
- Pk Info:ampoule of 1 ml