Supelco® Drug and metabolite standards, mycophenolic acid solution, (CRM)
Catalog # M-106-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:1,0 mg/ml in acetonitrile
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:24280-93-1
- Description:Mycophenolic acid solution
- Description:Mycophenolic acid solution
- Pk Info:ampoule of 1 ml