Supelco® Drug and metabolite standards, modafinil-d10 solution, (CRM)
M-103-1ML
: Merck
:
- Concentration:100 µg/ml in acetonitrile
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:1219804-30-4
- Description:Modafinil-d10 solution
- Description:Modafinil-d10 solution
- Pk Info:ampoule of 1 ml