Supelco® Drug and metabolite standards, metoprolol tartrate solution, (CRM)
Catalog # M-123-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:1,0 mg/ml in methanol (as free base)
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:56392-17-7
- Description:Metoprolol tartrate solution
- Description:Metoprolol tartrate solution
- Pk Info:ampoule of 1 ml