Supelco® Drug and metabolite standards, Fluoxetine-D6, (CRM)
Catalog # F-919-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 µg/ml in methanol (as free base)
- Pk:1 mL
- Standards:Certified reference material
- Description:Fluoxetine-D6
- Pk Info:ampoule of 1 ml