Supelco® Drug and metabolite standards, Ecgonine methyl ester solution, (CRM)
Catalog # E-001-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:1,0 mg/ml in acetonitrile
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:7143-09-1
- Description:Ecgonine methyl ester solution
- Description:Ecgonine methyl ester solution
- Pk Info:ampoule of 1 ml