Supelco® Drug and metabolite standards, Duloxetine hydrochloride solution, (CRM)
Catalog # D-044-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:1,0 mg/ml in methanol (as free base)
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:136434-34-9
- Description:Duloxetine hydrochloride solution
- Description:Duloxetine hydrochloride solution
- Pk Info:ampoule of 1 ml