Supelco® Drug and metabolite standards, Diltiazem hydrochloride solution, (CRM)
Catalog # D-035-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:1,0 mg/ml in acetonitrile (as free base)
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:33286-22-5
- Description:Diltiazem hydrochloride solution
- Description:Diltiazem hydrochloride solution
- Pk Info:ampoule of 1 ml