Supelco® Drug and metabolite standards, Dehydronorketamine hydrochloride solution, (CRM)
Catalog # D-046-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 µg/ml in acetonitrile
- Pk:1 mL
- Standards:Certified reference material
- Description:Dehydronorketamine hydrochloride solution
- Pk Info:ampoule of 1 ml