Supelco® Drug and metabolite standards, Benzodiazepine Multi-component Mixture-8 solution, (CRM)
Catalog # B-033-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:250 µg/ml each component in acetonitrile
- Pk:1 mL
- Standards:Certified reference material
- Description:Benzodiazepine Multi-component Mixture-8 solution
- Pk Info:ampoule of 1 ml