Supelco® Drug and metabolite standards, Atenolol solution, (CRM)
Catalog # A-072-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:1,0 mg/ml in acetonitrile
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:29122-68-7
- Description:Atenolol solution
- Description:Atenolol solution
- Pk Info:ampoule of 1 ml