Supelco® Drug and metabolite standards, Amitriptyline hydrochloride solution, (CRM)
Catalog # A-923-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:1,0 mg/ml in methanol
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:549-18-8
- Description:Amitriptyline hydrochloride solution
- Description:Amitriptyline hydrochloride solution
- Pk Info:ampoule of 1 ml