Supelco® Drug and metabolite standards, 4'-Hydroxydiclofenac solution, (CRM)
Catalog # H-052-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 µg/ml in acetonitrile
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:64118-84-9
- Description:4'-Hydroxydiclofenac solution
- Description:4'-Hydroxydiclofenac solution
- Pk Info:ampoule of 1 ml