Supelco® Cerilliant® Organic Reference Standard, 3,3′,5-Triiodo-L-thyronine-¹³C₆ solution
Catalog # T-077-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 µg/ml in methanol with 0,1 N NH₃
- Pk:1 mL
- Standards:Certified reference material, Cerilliant®
- CAS No.:1213431-76-5
- Description:3,3′,5-Triiodo-L-thyronine-¹³C₆ solution
- Storage conditions:–20 °C
- Hazard class description:H225 - H301 + H311 + H331 - H370
- Shipping conditions:WICE
- M-Clarity quality level:MQ300
- Molecular weight:656,93
- Flash point:9,7 °C
- Pk Info:Ampule of 1 ml
Specifications
About this item
- GHS signal: GHS02, GHS06, GHS08
Caution: P210 - P233 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311