Supelco® Cerilliant® Organic Reference Standard, 11-Deoxycortisol-D₅ (2,2,4,6,6-D₅)
D-078-1ML
: Merck
:
- Concentration:100 µg/ml in methanol
- Pk:1 mL
- Standards:Certified reference material, Cerilliant®
- CAS No.:1258063-56-7
- Description:11-Deoxycortisol-D₅ (2,2,4,6,6-D₅)
- Storage conditions:–20 °C
- Hazard class description:H225 - H301 + H311 + H331 - H370
- Shipping conditions:WICE
- MDL No.:MFCD15144809
- M-Clarity quality level:MQ300
- Synonym(s):11-Deoxycortisol-D5 (2,2,4,6,6-D5) solution; 11-Desoxycortisol-D₅ (2,2,4,6,6-D₅); Cortodoxone-D₅ (2,2,4,6,6-D₅)
- Molecular weight:351,49
- Flash point:9,7 °C - Closed cup
- Pk Info:Ampule of 1 ml
- GHS signal: GHS02, GHS06, GHS08
: P210 - P280 - P301 + P310 + P330 - P302 + P352 + P312 - P304 + P340 + P311