Drug and metabolite standards, (+/-)-Hydroxybupropion solutions, (CRM), Supelco®
Catalog # H-066-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:1,0 mg/ml in acetonitrile
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:92264-81-8
- Description:(+/-)-Hydroxybupropion solution
- Description:(+/-)-Hydroxybupropion solution
- Pk Info:ampoule of 1 ml