Drug and metabolite standards, (+/-)-cannabicyclol (CBL) solution, (CRM), Supelco®
C-154-1ML
: Merck
:
- Concentration:1,0 mg/ml in acetonitrile
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:21366-63-2
- Description:(+/-)-Cannabicyclol (CBL) solution
- Description:(+/-)-Cannabicyclol (CBL) solution
- Pk Info:ampoule of 1 ml