6-Chloro-3-indolyl phosphate p-toluidine salt (BCIP Pink)
Catalog # 10006.
Supplier: Biotium
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:354.73 g/mol
CAS Number:159954-33-3
Formula:C₈H₆ClNO₄P·C₇H₁₀N
MDL Number:MFCD00216690