


Cetrimonium bromide, powder
Catalog # ICNA0520823280
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Identity Test:Passes
- FTIR:Conforms to Standard
Specification Test Results
About this item
:
:364.45 g/mol
EINECS:200-311-3
:GHS05
:GHS07
:GHS09
Storage Temperature:Ambient
CAS Number:57-09-0
Formula:CH₃(CH₂)₁₅N(Br)(CH₃)₃
MDL Number:MFCD00011772