Supelco® Organic Reference Standard, Methyl Cis-11-Eicosenoate
Catalog # 17263-100MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle, bottomless
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:2390-09-2
- Description:Methyl cis-11-eicosenoate
- Storage conditions:–20 °C
- Shipping conditions:WICE
- MDL No.:MFCD00010458
- M-Clarity quality level:MQ100
- Synonym(s):cis-11-Eicosenoic acid methyl ester
- Formula:CH₃(CH₂)₇HC=CH(CH₂)₉CO₂CH₃
- Molecular weight:324,54
- Flash point:Not applicable