Bottles, stainless steel
Supplier: Sampling Systems
Certificates
About this item
Stainless steel bottles are ideal for storing and transporting powders and liquids. The high-quality, crevice-free finish makes these containers ideal for the most demanding applications.
- 316L stainless steel body and lid
- Multi-purpose containers
- Robust construction
- All welds are ground and polished smooth so its ideal for hygienic industries
- Fitted with a 2" tri-ferrule leakproof lid system
The containers come as standard with a removable, easy clean silicone seal.
Certifications: Seal is food grade and meets all the latest FDA and EC compliance standards.
Delivery: Supplied with lid, silicone gasket (FDA compliant), and clampband.
Frequently Bought Together