Disinfectant, non sterile, Incidin® Pro
Catalog # SHIE3076720
Supplier: ECOLAB CONTAMINATION CONTROL
Specifications
- Description:Incidin® pro, 1×6,0 l
- Packed:Canister
- Pk:1
Specifications
About this item
Incidin® Pro provides a broad spectrum efficacy combined with excellent material compatibility.
- Fragrance and aldehyde free
- Short contact time
Certifications: Bactericidal: EN 1276, EN 13697; Yeasticidal: EN 1650, EN 13697.
Caution: Use biocides safely. Always read the label and product information before use.