TLC spray box
Catalog # S1509-5EA
Supplier: Merck
Specifications
- Description:TLC spray box
- Pk:1
Specifications
About this item
This TLC spray box may be used to spray reagent for TLC plates. Can be used in thin layer chromatography and HPTLC.
The reagent/chemical spraying on the TLC plates are generally carried out inside a fume hood or commercially available TLC spray cabinet with blower and exhaust hose. The TLC plate can be placed supported upright or on a paper inside the spray box. Spraying the box ensures complete removal of reagent mist.
Compatible with most Sigma sprays.