Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Trays/Dishes/Pots
Instrument Trays/Dishes
Instrument tray
Instrument tray
Instrument tray
Catalog # 216-0486
Supplier:
Azpack
Expand image
Instrument tray
Catalog # 216-0486
Supplier:
Azpack
Specifications
Colour:
Translucent white
W×D×H:
300×250×50 mm
Pk:
1
Specifications
About this item
Product Details
PP, translucent white.
Product Details