Order Entry
Northern Ireland
ContactUsLinkComponent
 
Zinc sulphate heptahydrate 99.0-103.0%, Puriss., meets analytical specification of BP, Ph. Eur., USP, FCC, Fluka™
  14455-1KG
 :  
Zinc sulphate heptahydrate 99.0-103.0%, Puriss., meets analytical specification of BP, Ph. Eur., USP, FCC, Fluka™
  14455-1KG
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg
  • Seq:
    4024
  • Environmentally Preferable:

 

 

  • Assay:
    99.0 - 103.0 %
  • pH (5 %, 20°C):
    4.4 - 5.6
  • Arsenic (As):
    Max. 2 ppm
  • Calcium (Ca):
    Max. 0.005 %
  • Cadmium (Cd):
    Max. 2 ppm
  • Iron (Fe):
    Max. 10 ppm
  • Mercury (Hg):
    Max. 2 ppm
  • Potassium (K):
    Max. 0.01 %
  • Magnesium (Mg):
    Max. 0.005 %
  • Sodium (Na):
    Max. 0.01 %
  • Lead (Pb):
    Max. 4 ppm
  • Selenium (Se):
    Max. 0.001 %
  • Chloride (Cl):
    Max. 50 ppm
  • Appearance of the solution:
    complying
  • Alkalis/earth alkalis (as SO₄):
    Max. 0.5 %
  • Acidity:
    complying
  • Residual solvents:
    complying

 

 

 :
 :287.54 g/mol
 :100 °C
 :231-793-3
 :GHS07
 :GHS05
 :Ambient
 :7446-20-0
 :ZnSO₄·7H₂O
 :MFCD00149894
 :1.96 g/cm³ (20 °C)
 :12,10293