Nickel(II) chloride hexahydrate ≥98%, Puriss. p.a., Fluka™
Catalog # 31462-1KG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Assay:Min. 98.0 %
- pH (5 %, 20°C):4-7
- Calcium (Ca):Max. 0.005 %
- Cadmium (Cd):Max. 0.001 %
- Cobalt (Co):Max. 0.01 %
- Copper (Cu):Max. 0.001 %
- Iron (Fe):Max. 0.002 %
- Magnesium (Mg):Max. 0.001 %
- Sodium (Na):Max. 0.005 %
- Lead (Pb):Max. 0.001 %
- Zinc (Zn):Max. 0.001 %
- Sulfate (SO₄):Max. 0.005 %
Specification Test Results
About this item
:
:237.69 g/mol
Merck Index:13,06529
Melting Pt:80 °C
Density:3,550 g/cm³ (20 °C)
UN:3288
ADR:6.1,III
:GHS08
:GHS06
Storage Temperature:Ambient
CAS Number:7791-20-0
Formula:NiCl₂·6H₂O
MDL Number:MFCD00149809