


1,2-Dimethoxyethane ≥99% inhibitor-free, Reagent Grade
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:110-71-4
:
Formula:C₄H₁₀O₂
MDL Number:MFCD00008502
:90.12 g/mol
Merck Index:13,03251
Boiling Pt:85 °C (1,013 hPa)
Melting Pt:–58 °C
Density:0,87 g/cm³ (20 °C)
Flash Pt:–2 °C
Storage Temperature:Ambient
EINECS:203-794-9
UN:2252
ADR:3,II
:GHS02
:GHS08
:GHS07
Specification Test Results
- Identity (IR):complying
- Assay (GC):Min. 99.0 %
- Water (Karl Fischer):Max. 0.5 %
- Peroxides (as H₂O₂):Max. 15 ppm
- Vapor Pressure:48 mmHg ( 20 °C)
- Explosion Limit:10.4 %
Specification Test Results
Frequently Bought Together