

Sodium nitrate 99.0-100.5%, meets analytical specification of FCC, E251, Fluka™
13444-1KG
:
- Pack type:Plastic bottle
- Pk:1 kg
- Seq:4024
- Assay (calculated to the dried substance):99.0 - 100.5 %
- Loss on drying (105°C):Max. 1 %
- pH (5 %, 20°C):5-7
- Arsenic (As):Max. 2 ppm
- Calcium (Ca):Max. 0.005 %
- Copper (Cu):Max. 0.001 %
- Iron (Fe):Max. 0.001 %
- Mercury (Hg):Max. 1 ppm
- Potassium (K):Max. 0.01 %
- Magnesium (Mg):Max. 0.002 %
- Lead (Pb):Max. 2 ppm
- Zinc (Zn):Max. 0.001 %
- Heavy metals (as Pb):Max. 0.001 %
- Chloride (Cl):Max. 0.001 %
- Nitrite (NO₂):Max. 10 ppm
- Sulfate (SO₄):Max. 0.01 %
- Total halogen compounds (as Cl):Max. 0.02 %
- Residual solvents:complying
:
:84.99 g/mol
:13,08720
:Decomposes on heating
: Dec. (on heating)
:308 °C
:2,26 g/cm³ (20 °C)
:Ambient
:231-554-3
:1498
:5.1,III
:01-2119488221-41
:GHS07
:GHS03
:7631-99-4
:NaNO₃
:MFCD00011119