


Methylcyclohexane 99%, Reagent Grade
:108-87-2
:
:C₆H₁₁CH₃
:MFCD00001497
:98.19 g/mol
:13,06074
:6,7 %(V)
:–126 °C
:0,769 g/cm³ (20 °C)
:–4 °C
:Ambient
:203-624-3
:2296
:3,II
:GHS07
:GHS08
:GHS02
- Identity (IR):complying
- Assay (GC):Min. 98.5 %
- Vapor Pressure:83.2 mmHg ( 37.7 °C)
- Refractive Index:n20/D 1.422 (lit.)
- Vapor Density:3.4 (vs air)
- Autoignition Temperature:545 °F
Frequently Bought Together