Order Entry
Northern Ireland
ContactUsLinkComponent
 
 
 
Dichloromethane ≥99.9% stabilised, Biotechnology Grade, Burdick & Jackson™
 
undefined
 
 
Dichloromethane ≥99.9% stabilised, Biotechnology Grade, Burdick & Jackson™

 

  • Identity (IR):
    complying
  • Assay (GC):
    Min. 99.85 %
  • Non-volatile matter:
    Max. 0.0003 %
  • Water (Karl Fischer):
    Max. 0.002 %
  • 2-methyl-2-buten (GC):
    40 - 150 ppm
  • Absorbance at 400 nm:
    Max. 0.01
  • Absorbance at 340 nm:
    Max. 0.01
  • Absorbance at 260 nm:
    Max. 0.02
  • Absorbance at 250 nm:
    Max. 0.05
  • Absorbance at 240 nm:
    Max. 0.20
  • Absorbance at 235 nm:
    Max. 1.0
  • Vapor Density:
    2.9 (vs air)
  • Refractive Index:
    n20/D 1.424 (lit.)
  • Contains:
    40-150 ppm amylene as stabilizer
  • Explosion Limit:
    22 %

 

 

Frequently Bought Together