

Cyclopentyl methyl ether (CPME), EMPLURA®
Supplier: Merck
Certificates
About this item
CAS Number:5614-37-9
:
Formula:C₆H₁₂O
MDL Number:MFCD08276401
:100.16 g/mol
Melting Pt:–140 °C
Density:0,86 g/cm3 (20 °C)
Flash Pt:–1 °C
UN:3271
ADR:3,II
:M-Clarity™ quality level = MQ200
:GHS02
:GHS07
Limited Time Sale
Select items below are currently on sale
Frequently Bought Together