Order Entry
Northern Ireland
ContactUsLinkComponent
Sodium hexametaphosphate, J.T.Baker®
Sodium hexametaphosphate, J.T.Baker®
Catalog # V030-09
Supplier:  Avantor
CAS Number:  
undefined
Sodium hexametaphosphate, J.T.Baker®
Catalog # V030-09
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Poly bottle, white, blue T/E cap
  • Pk:
    3 kg
  • Seq:
    4090

Specifications

Specification Test Results

  • Assay (as P₂O₅):
    ≥65.0 %

Specification Test Results

About this item

 :
 :611.77 g/mol
Melting Pt:610 °C
Density:1,25 g/cm³ (20 °C)
EINECS:233-343-1
CAS Number:10124-56-8
 :68915-31-1
Formula:Na₆P₆O₁₈
MDL Number:MFCD00136045