Analytical standard, Oxyclozanide-(benzoyl ring-¹³C₆), reference material, Honeywell Fluka™
Catalog # HONCR1377-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:1353867-74-9
- Description:Oxyclozanide-(benzoyl ring-¹³C₆)
- Melting point:205…210 °C
- Formula:¹³C₆C₇H₆Cl₅NO₃
- Storage temperature:Ambient
- Description:Oxyclozanide-(benzoyl ring-¹³C₆)
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results