Analytical standard, Benalaxyl-M, reference material, Honeywell Fluka™
Catalog # HONCR1363-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:98243-83-5
- Description:Benalaxyl-M
- Melting point:71…73 °C
- Formula:C₂₀H₂₃NO₃
- Storage temperature:Ambient
- Description:Benalaxyl-M
- Boiling point:280…290 °C
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results