Analytical standard, Cimbuterol, reference material, Honeywell Fluka™
Catalog # HONCR1352-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:54239-39-3
- Description:Cimbuterol
- Melting point:143.2 °C
- Formula:C₁₃H₁₉N₃O
- Storage temperature:Ambient
- Description:Cimbuterol
- Boiling point:438.7 °C
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results