Analytical standard, Selamectin, reference material, Honeywell Fluka™
Catalog # HONCR1341-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:220119-17-5
- Description:Selamectin
- Formula:C₄₃H₆₃NO₁₁
- Storage temperature:2…8 °C
- Description:Selamectin
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results