Analytical standard, TEPP, reference material, Honeywell Fluka™
Catalog # HONCR1338-50MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:50 mg
- Standards:Analytical standard, neat
- CAS No.:107-49-3
- Description:TEPP
- Formula:C₈H₂₀O₇P₂
- Storage temperature:2…8 °C
- Description:TEPP
- Boiling point:140…142 °C
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results