Order Entry
Northern Ireland
ContactUsLinkComponent
 
Analytical standard, Meperfluthrin, reference material, Honeywell Fluka™
  HONCR1313-25MG
 :  
Analytical standard, Meperfluthrin, reference material, Honeywell Fluka™
  HONCR1313-25MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    25 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    915288-13-0
  • Description:
    Meperfluthrin
  • Formula:
    C₁₇H₁₆Cl₂F₄O₃
  • Storage temperature:
    Ambient
  • Description:
    Meperfluthrin

 

 

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results