Analytical standard, Linuron-(methyl-d₃, methoxy-d₃), reference material, Honeywell Fluka™
Catalog # HONCR1297-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:1219804-76-8
- Description:Linuron-(methyl-d₃, methoxy-d₃)
- Formula:C₉D₆H₄Cl₂N₂O₂
- Storage temperature:Ambient
- Description:Linuron-(methyl-d₃, methoxy-d₃)
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results