Order Entry
Northern Ireland
ContactUsLinkComponent
 
Analytical standard, Monoquat iodide-(methyl-d₃), reference material, Honeywell Fluka™
Catalog # HONCR1294-25MG
CAS Number:  
Analytical standard, Monoquat iodide-(methyl-d₃), reference material, Honeywell Fluka™
Catalog # HONCR1294-25MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    25 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    22884-38-4
  • Description:
    Monoquat iodide-(methyl-d₃)
  • Melting point:
    249…252 °C
  • Formula:
    C₁₁D₃H₈IN₂
  • Storage temperature:
    Ambient
  • Description:
    Monoquat iodide-(methyl-d₃)

Specifications

About this item

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results