Analytical standard, Methoxychlor-d14 (bis(4-methoxyphenyl-d₇)), reference material, Honeywell Fluka™
Catalog # HONCR1286-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:1644449-82-0
- Description:Methoxychlor-d14 (bis(4-methoxyphenyl-d₇))
- Melting point:74…76 °C
- Formula:C₁₆D₁₄HCl₃O₂
- Storage temperature:Ambient
- Description:Methoxychlor-d14 (bis(4-methoxyphenyl-d₇))
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results