Analytical standard, Chlorthiophos-sulfone, reference material, Honeywell Fluka™
Catalog # HONCR1283-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:25900-20-3
- Description:Chlorthiophos-sulfone
- Melting point:72…76 °C
- Formula:C₁₁H₁₅Cl₂O₅PS₂
- Storage temperature:2…8 °C
- Description:Chlorthiophos-sulfone
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results