Analytical standard, Carbaryl-(methyl-d₃), reference material, Honeywell Fluka™
HONCR1273-10MG
:
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:1433961-56-8
- Description:Carbaryl-(methyl-d₃)
- Melting point:128…131 °C
- Formula:C₁₂D₃H₈NO₂
- Storage temperature:Ambient
- Description:Carbaryl-(methyl-d₃)
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results