Order Entry
Northern Ireland
ContactUsLinkComponent
Analytical standard, Terbutaline acetate salt hemihydrate, reference material, Honeywell Fluka™
Analytical standard, Terbutaline acetate salt hemihydrate, reference material, Honeywell Fluka™
Catalog # HONCR1219-10MG
CAS Number:  
Analytical standard, Terbutaline acetate salt hemihydrate, reference material, Honeywell Fluka™
Catalog # HONCR1219-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • Description:
    Terbutaline acetate salt hemihydrate
  • Melting point:
    97 °C
  • Formula:
    C12H19NO3 · C2H4O2 · 0.5H2O
  • Storage temperature:
    Ambient
  • Description:
    Terbutaline acetate salt hemihydrate

Specifications

About this item

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results