Order Entry
Northern Ireland
ContactUsLinkComponent
 
Analytical standard, Cafenstrole, reference material, Honeywell Fluka™
  HONCR1163-50MG
 :  
Analytical standard, Cafenstrole, reference material, Honeywell Fluka™
  HONCR1163-50MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    50 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    125306-83-4
  • Description:
    Cafenstrole
  • Melting point:
    114…116 °C
  • Formula:
    C₁₆H₂₂N₄O₃S
  • Storage temperature:
    Ambient
  • Description:
    Cafenstrole

 

 

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results