Analytical standards, Parabens, reference material, Honeywell Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Melting point:–95 °C
- Storage temperature:2…8 °C
- Description:Paraben target analyte mix solution
- Boiling point:55…57 °C
Specifications
Frequently Bought Together