Analytical standard, Flupyrsulfuron-methyl sodium, reference material, Honeywell Fluka™
Catalog # HONCR1068-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:144740-54-5
- Description:Flupyrsulfuron-methyl sodium
- Formula:C₁₅H₁₃F₃N₅NaO₇S
- Storage temperature:Ambient
- Description:Flupyrsulfuron-methyl sodium
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results