Order Entry
Northern Ireland
ContactUsLinkComponent
Analytical standard, Trietazine-(monoethyl-d₅-amino), reference material, Honeywell Fluka™
Analytical standard, Trietazine-(monoethyl-d₅-amino), reference material, Honeywell Fluka™
  HONCR1046-10MG
 :  
Analytical standard, Trietazine-(monoethyl-d₅-amino), reference material, Honeywell Fluka™
  HONCR1046-10MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    1397243-73-0
  • Description:
    Trietazine-(monoethyl-d₅-amino)
  • Formula:
    C₉D₅H₁₁ClN₅
  • Storage temperature:
    Ambient
  • Description:
    Trietazine-(monoethyl-d₅-amino)

 

 

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results