Analytical standard, Fenbendazole sulfone, reference material, Honeywell Fluka™
Catalog # HONCR1035-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:54029-20-8
- Description:Fenbendazole sulfone
- Melting point:320 °C
- Formula:C₁₅H₁₃N₃O₄S
- Storage temperature:Ambient
- Description:Fenbendazole sulfone
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results